Learn More
Research Products International Corp WorkStation Lab Mat, Purple
SDP
Supplier:  Research Products International Corp 212618P
Encompass
Reusable Workstation Lab Mat is ideal for keeping laboratory benchtop organized and workflow processes optimized. Silicone surface features grid surface on one side for tube staging and prevents items from moving. Maintains clean benchtop, prevents stains, and spills. Anti-skid surface helps keep items in place, reduces vibrational sound, and dampens noise from equipment. Personal size mat is easy to transport and allows user to easily create and protect work area. Silicone mat is FDA food grade, chemically resistant, durable, washable, and stain resistant. Lip on edge can collect up to 250ml of spilled liquid.
Keeps benchtop organized
Easy clean up
Anti-skid, grid surface
Catalog No. 50-222-9639