Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/4 ID PK10
Supplier:  Grainger 16P299
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel from Grainger is a worm gear style hose clamp made from 201 stainless steel. It has a band width of 7/16", a clamping range of 5/8" to 1-1/4", and a screw size of 5/16"-18. This hose clamp is designed for automotive and industrial applications. The 201 stainless steel offers good corrosion resistance. The worm gear mechanism allows for a wide clamping range while maintaining high clamping force on hoses and fittings. This is a high quality stainless steel hose clamp well-suited for many applications.
Catalog No. 50-276-7384