Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/2 ID PK10
Supplier:  Grainger 16P301
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel is a stainless steel worm gear clamp with a 201 stainless steel band. It has a worm screw drive for tightening and is designed for heavy duty applications. This hose clamp has a band width of 7/8", a band thickness of 0.050", and a clamping range of 1-1/4" to 2". It can provide up to 750 lbs of clamping force. The 201 stainless steel offers good corrosion resistance. This heavy duty worm gear clamp is ideal for automotive and industrial hose connections.
Catalog No. 50-276-7353