Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/4 ID PK10
Supplier:  Grainger 16P278
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/2 inch. The clamp features a rolled edge, screw housing, and worm drive adjusting screw. It is designed for automotive and industrial hose connections. This stainless steel worm gear clamp provides good corrosion resistance and durability.
Catalog No. 50-276-5158