Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 3/4 ID PK10
Supplier:  Grainger 16P306
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1-1/4 inches to 2 inches. This hose clamp creates a constant radial pressure and allows single hand tightening. It is designed for automotive and industrial applications. The 201 stainless steel offers good corrosion resistance. This is a high quality worm drive hose clamp suitable for a variety of hose connections.
Catalog No. 50-276-7670