Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 ID PK10
Supplier:  Grainger 16P303
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2" and a clamping range of 1-1/4" to 2". The clamp has a screw housing made of glass-filled nylon with a worm drive screw of hardened steel. It is designed for automotive and industrial hose connections. The 201 stainless steel provides good corrosion resistance. This is a heavy duty worm drive clamp suitable for high vibration applications.
Catalog No. 50-276-7354