Learn More
Grainger Worm Gear Hose Clamp 1/2 W 2 1/2 ID PK10
Supplier:  Grainger 16P286
Encompass_Preferred
The Grainger 2-1/2" to 4-1/2" SAE 64 Hose Clamp is a worm-drive hose clamp made from 201 stainless steel banding with a 201 stainless steel screw. It has a band width of 15/32" and a clamping range of 2-1/2" to 4-1/2". This hose clamp is sold in a pack of 10 pieces. It is designed for automotive and industrial hose connections. The clamp has an Intertek ETL Listed mark for quality assurance.
Catalog No. 50-276-5648