Learn More
Grainger Worm Gear Hose Clamp 1/2 W 3 1/2 ID PK10
Supplier:  Grainger 16P311
Encompass_Preferred
The Worm Gear Hose Clamp 301 Stainless Steel by Oetiker is a worm drive hose clamp made from 301 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 inches to 1-1/4 inches. This hose clamp features a rolled edge, screw housing and worm drive adjusting screw. It is designed for automotive and industrial hose connections. The 301 stainless steel provides good corrosion resistance. This is a high quality worm gear clamp suitable for a wide range of hose sizes.
Catalog No. 50-276-8301