Learn More
Grainger Worm Gear Hose Clamp 1/2 W 3 ID PK10
Supplier:  Grainger 16P310
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1-1/4 inches to 2 inches. This hose clamp creates a constant radial pressure to prevent leaks. It is recommended for automotive and industrial applications. The clamp is made in Switzerland and offers corrosion resistance.
Catalog No. 50-276-8367