Learn More
Grainger Worm Gear Hose Clamp 5/8 W 2 1/4 ID PK10
Supplier:  Grainger 16P221
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a 304 stainless steel worm gear hose clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a working range of 5/8 inch to 1-1/4 inch diameter. It is made in the USA and conforms to ASME B16.20 and SAE J1508 standards. The clamp is available in pack sizes of 10, 25, 50 or 100 pieces.
Catalog No. 50-276-4560