Learn More
Grainger Worm Gear Hose Clamp 5/8 W 2 1/4 ID PK10
Supplier:  Grainger 16P237
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/4 inch. This hose clamp features a 201 stainless steel band and housing for corrosion resistance. The worm drive allows the clamp to be tightened by turning the screw to provide up to 3000 psi of clamping force. It can be used for automotive, industrial, and marine applications. The clamp meets SAE J1508 Type 201 standards.
Catalog No. 50-276-3712