Learn More
Sigma Aldrich Fine Chemicals Biosciences Worthington LS Series Liqu1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7439981EA
Encompass
These Dewars with canister and rack storage systems offer long-term cost-effective storage for cells tissues and other biological samples. Canisters hold samples in either the vapor or liquid phase depending on the liquid nitrogen level. Temperatures are maintained from -184C under the closed necktube core to -196C at the liquid nitrogen level..nbspRibbed high strength aluminum body magneformed necktube design and durable paint system.nbspRack index location ring and internal location fixture computer compatible box storage is ideal for simple inventory management.nbspSuperior vacuum performance.nbspLockable lids
Catalog No. 50-290-8636
Add to Cart
Request a Quote