Learn More
Cayman Chemical Wortmannin, 19545-26-7, 1 mg
SDP
Supplier:  Cayman Chemical 100105911
Encompass_Preferred
Molecular Formula C23H24O8, Purity ≥98%, Formula Weight 428.4, Synonyms: KY 12420; A potent, cell-permeable, and irreversible inhibitor of PI3K enzymes (IC50 = 1-10 nM).
Catalog No. NC0744498
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.