Learn More
Apexbio Technology LLC Wortmannin, 20mg. Cas: 19545-26-7 MFCD: MFCD00133927. PI3K inhibitor,selective and irreversible
SDP
Supplier:  Apexbio Technology LLC A854420
Encompass_Preferred
Wortmannin is a selective and irreversible inhibitor of phosphatidylinositol-3-kinase with IC50 value of 1.9nM [1].Wortmannin is a microbial product. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5159