Learn More
Medchemexpress LLC HY-10197 25mg , Wortmannin SL-2052;KY-12420 CAS:19545-26-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1019725MG
Encompass_Preferred
Medchemexpress, HY-10197 25mg Wortmannin SL-2052;KY-12420 CAS:19545-26-7 Formula:C23H24O8 IC50: 3 nM (PI3K), 200 nM (MLCK) IC50: 16 nM (DNA-PK), 150 nM (ATM), 1.8 μM (ATR) Purity:98% Wortmannin is a multi-target inhibitor of PI3K and MLCK with IC 50 s of 3 nM and 200 nM, respectively. Wortmannin is also a potent inhibitor of DNA-PK ( IC 50 , 16 nM) and ATM (IC 50 , 150 nM). Wortmannin is also a potent inhibitor of Polo-like kinase ( Plk ). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3996