Learn More
Medchemexpress LLC WP1066 | 857064-38-1 | C17H14BrN3O | 5 MG

Supplier: Medchemexpress LLC HY153125MG
WP1066 is an inhibitor of JAK2 and STAT3, also demonstrating effects on STAT5 and ERK1/2, without impacting JAK1 and JAK3. This compound can cross the blood-brain barrier and is intended for research use only.
- Purity of 99.77%
- Appearance as a white to light yellow solid
- Targets JAK2 and STAT3
- Affects STAT5 and ERK1/2
- No impact on JAK1 and JAK3
- Crosses the blood-brain barrier
- Soluble in DMSO and ethanol
- Inhibits the growth of various cancer cell lines
- Enhances the cytotoxic effects of CTX on tumors
- Excellent CNS penetration for potential cancer research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.