Learn More
Selleck Chemical LLC Degrasyn (WP1130) 50mg 856243-80-6
SDP
Supplier:  Selleck Chemical LLC S224350MG
Encompass_Preferred
Degrasyn (WP1130) is a selective deubiquitinase (DUB: USP5, UCH-L1, USP9x, USP14, and UCH37) inhibitor and also suppresses Bcr/Abl, also a JAK2 transducer (without affecting 20S proteasome) and activator of transcription (STAT). Degrasyn (WP1130) induces apoptosis and blocks autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-807-4