Learn More
World Precision Instrument WPI Swiss Scissors, 9 cm, Curved, Fine Blunt Tip
SDP
Supplier:  World Precision Instrument 504615
Encompass
Swiss microdissecting scissors are ring handled surgical scissors particularly suited for dissection and surgical tasks that require precision cutting. These scissors are crafted with high-quality materials and are known for their durability and sharpness. Swiss-made surgical instruments are renowned for their precision and craftsmanship. Swiss manufacturers adhere to strict quality control standards, and their products are known for precision and reliability. WPI Swiss dissecting scissors are made from high-grade stainless steel, which provides corrosion resistance and ensures a long lifespan for the instrument. All these scissors are 9 cm (3.5”) long. These scissors have a fine, slender design with sharp blades, allowing for precise cutting in delicate procedures. They have sharp, blunt or rounded tips These are available with curved or straight tips. Extra fine, fine, and heavy-duty blades are available. You can also get the unique beveled tip design.
Catalog No. 50-253-9327