Learn More
Med Vet International Cloth Instrument Wrap, 12" x 12", 5/pack
Supplier:  Med Vet International W1212X5
Encompass_Preferred
Autoclavable Wraps are a woven blend of 55% cotton and 45% medium weight polyester, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability These two-ply wraps are stitched with a bias binding that is also color-coded for quick identification
Catalog No. 50-304-4516
 Restricted Item