Learn More
Med Vet International Quick Check* H400 Sterilization Wrap White / Blue 36 X 36 inch Dual Layer Sms Polypropylene Steam / Eo Gas / Hydrogen Peroxide, 34158, 72/Case
Supplier:  Med Vet International 809429CS
Encompass_Preferred
Comes with white and blue layers that are bonded together – inspect trays easier and faster than ever Superior microbial barrier protection with patented Powerguard technology Use with most basin or instrument sets, procedure trays, drills Made from recyclable number 5 polypropylene fabric There are 72 3-by-3-feet sterilization wraps in a case
Catalog No. 50-282-9650