Learn More
Accuflow Systems 2 pc 3/8” & 1/2” Dr. Crowfoot Spring Wrench Set
Supplier:  Accuflow Systems 58403
Encompass_Preferred
The Spring Jaw Crowfoot wrench has a unique design has endless applications from bolt and screw extraction to loosening and tightening hard-to-reach pipes and fittings. The dual connector allows you to connect a ratchet or breaker bar on the bottom or side, taking full advantage of the ratcheting function and gripping teeth. The ratcheting action design allows the tool to rotate back and reposition, allowing for better performance than regular crowfoot wrenches, especially in tight spaces. The serrated gripping teeth with spring-loaded, self-adjusting jaws firmly grip all type of fasteners including hexagonal, spline, star & square head nuts, bolts & screws, including damaged fasteners. Use with breaker bars, ratchets & extension bars. Set has a lifetime warranty.Set Includes:3/8″ Dr. & 1/2″ Dr. Crowfoot3/8″ Dr. Size Range: 8mm – 17mm (5/16″ – 11/16″)1/2″ Dr. Size Range: 14mm – 32mm (9/16″ – 1-1/4″)
Catalog No. 50-223-6804