Learn More
Selleck Chemical LLC WRW4 S9818-5MG
Supplier: Selleck Chemical LLC S98185MG
WRW4 is a specific formyl peptide receptor-like 1 (FPRL1) antagonist that inhibits WKYMVm binding to FPRL1 with IC50 of 0 23 M WRW4 is a selective antagonist of formyl peptide receptor type 2 (FPR2)/Lipoxin A4 receptor (ALX)