Learn More
Medchemexpress LLC WSB1 degrader 1 | 2306039-66-5 | 99.8% | C21H22N2O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14148210MG
Encompass_Preferred
WSB1 Degrader 1 is a small-molecule, orally active degrader that targets WSB1 (WD repeat and SOCS box-containing 1). It induces WSB1 degradation in a time- and dose-dependent manner and has shown anti-metastatic activity in cellular assays and in mouse models, making it suitable for preclinical studies of metastasis and WSB1 biology.
  • Induces WSB1 degradation in cells.
  • Demonstrated antimigration activity in multiple cancer cell lines.
  • Reduces pulmonary metastasis in mouse models.
  • Available as solid or as a 10 mM solution in DMSO for experimental flexibility.
  • Molecular formula C21H22N2O2 and molecular weight 334.41.
  • High reported purity (≈99.8%) suitable for research applications.
Catalog No. 50-002-58417