Learn More
Medchemexpress LLC WSB1 Degrader 1 | 2306039-66-5 | MFCD34368539 | 99.8% | 334.41 g·mol⁻1 | C21H22N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1414825MG
Encompass_Preferred
WSB1 Degrader 1 is a small-molecule research degrader that targets WSB1 (WD repeat and SOCS box-containing 1). It is described as potent and orally active with demonstrated anticancer and anti-metastatic effects, intended for biochemical, cellular, and early in vivo studies.
  • Potent degrader of WSB1 with reported anticancer and anti-metastatic effects.
  • Orally active in preclinical studies, suitable for in vivo research.
  • High purity (99.81%), appropriate for sensitive assays.
  • Good solubility in DMSO (25 mg/mL) and formulatable for in vivo use.
  • Stable when stored as a powder under recommended conditions.
  • Available in small research pack sizes for early-stage experiments.
Catalog No. 50-002-22266