Learn More
Medchemexpress LLC HY-136976 100mg , WST-1 CAS:150849-52-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136976100MG
Encompass_Preferred
HY-136976 100mg WST-1 CAS: 150849-52-8 WST-1 is a Tetrazolium dye, widely used in the cell viability assays. WST-1 produces a water-soluble formazan product (WST-1 formazan) that occurs at the cell surface in NADPH dependent manner in viable cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0091