Learn More
Selleck Chemical LLC WST-8-25mg
SDP
Supplier:  Selleck Chemical LLC S776425MG
Encompass_Preferred
WST-8 is a water-soluble tetrazolium salt used for assessing cell metabolic activity that produces corresponding formazan dye that absorbs at 460 nm. WST-8 is typically used as a cell viability indicator in cell proliferation assays.
Catalog No. 50-238-9606