Learn More
Fondriest Environmental Inc WT 25 Rhodamine WT dye, 2.5% active ingredient, 1 pint
SDP
Supplier:  Fondriest Environmental Inc 10602301P
Encompass
The Bright Dyes Fluorescent FWT red dye is ideal for septic inspection applications. Preferred, high strength formulations for medium to large scale visual and fluoremetric studies. NSF Standard 60 Certification for use in or around potable water sources. Also used to calibrate YSI 6025, 6130, 6131 & 6132 optical sensors.
Catalog No. NC1649301