Learn More
Selleck Chemical LLC WY-14643
SDP
Supplier:  Selleck Chemical LLC S802950MG
Encompass_Preferred
WY-14643 (Pirinixic Acid NSC 310038) is a potent and selective PPAR activator with an EC50 of 1 5 M
Catalog No. 50-000-04705