Learn More
Enzo Life Sciences WY-14643 (250mg). CAS: 50892-23-4
SDP
Supplier:  Enzo Life Sciences BMLGR2000250
Encompass_Preferred
WY-14643 is a potent and widely used peroxisome proliferator activator receptor (PPAR) activator. It has been shown to be selective for PPARα. Alternative name: Pirinixic acid. Purity: ≥97% (HPLC). Solubility: Soluble in ethyl acetate or dilute base or slightly soluble in alcohol. Long Term Storage: Ambient.
Catalog No. 50-201-0927