Learn More
Medchemexpress LLC Wyc-209 | 2131803-90-0 | MFCD32067940 | 99.8% | 368.45 | C20H20N2O3S | 100MG

Supplier: Medchemexpress LLC HY124136100MG
WYC-209 is a synthetic retinoid research compound that functions as a retinoic acid receptor (RAR) agonist. It has been reported to induce apoptosis in tumor-repopulating cells via the caspase-3 pathway and is supplied as a powder for laboratory research.
- Acts as a retinoic acid receptor agonist.
- Induces apoptosis via caspase-3 (reported IC50 ≈ 0.19 μM).
- Molecular formula: C20H20N2O3S.
- Molecular weight: 368.45 g/mol.
- Purity: 99.8% (as supplied).
- Available as powder in pack sizes including 100 mg.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years).
- For research use only; not for human or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.