Learn More
TARGETMOL CHEMICALS INC WYE-354 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T673125MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WYE-354(IC50=5 nM) is an effective selective and ATP-competitive mTOR inhibitor. It blocks mTORC2/P-AKT(S473) and mTORC1/P-S6K(T389) not P-AKT(T308). The selectivity for mTOR is higher than PI3K(alpha) (>100-fold) and PI3K(gamma) (>500-fold). purity: 99%
Catalog No. 50-257-9298