Learn More
Medchemexpress LLC Wyrgrl | 1021155-39-4 | 99.9% | 849.98 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP10739
Encompass_Preferred
WYRGRL is a collagen II-targeting peptide designed to bind specifically to collagen II α1. This short peptide sequence is effective in cartilage targeting and has been shown to significantly enhance the cartilage-targeting efficiency of peptide-functionalized nanoplatforms in vivo.
  • Binds to collagen II α1
  • Short cartilage-targeting peptide sequence
  • Increases cartilage-targeting efficiency for peptide-functionalized nanoplatforms in vivo by approximately 72-fold
  • For research use only
  • Available in multiple sizes
  • Stable under recommended storage conditions
  • Appears as a white to off-white solid
Catalog No. 50-004-24894