Learn More
Apexbio Technology LLC WZ4003, 50mg. Cas: 1214265-58-3 MFCD: MFCD28015100. NUAK1/2 inhibitor, potent and selective
SDP
Supplier:  Apexbio Technology LLC B137450
Encompass_Preferred
WZ4003 is a potent and selective inhibitor of NUAK1 and NUAK2 with IC50 values of 20 and 100 nM, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5224