Learn More
Selleck Chemical LLC WZ4003 5mg 1214265-58-3
SDP
Supplier:  Selleck Chemical LLC S73175MG
Encompass_Preferred
WZ4003 is a highly specific NUAK kinase inhibitor with IC50 of 20 nM and 100 nM for NUAK1 and NUAK2 in cell-base assays, respectively, without significant inhibition on 139 other kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4637