Learn More
Medchemexpress LLC HY-19331 10mg , WZB117 CAS:1223397-11-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1933110MG
Encompass_Preferred
HY-19331 10mg WZB117 CAS: 1223397-11-2 WZB117 is a glucose transporter 1 (Glut1) inhibitor, which downregulates glycolysis, induces cell-cycle arrest, and inhibits cancer cell growth in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1035