Learn More
Medchemexpress LLC X-GalNAc 25mg
SDP
Supplier:  Medchemexpress LLC 25MGHYW116594
Encompass_Preferred
X-GalNAc is a chromogenic substrate for for N-acetyl- -galactosidase used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates[1]
Catalog No. 50-002-05858