Learn More
Alkali Scientific X GlcA Cyclohexylammonium Salt [5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt], 1 G
SDP
Supplier:  Alkali Scientific XGL004CP
Encompass_Preferred
X GlcA Cyclohexylammonium Salt (5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt) is a chromogenic substrate used in biochemical research and diagnostic assays. This 1 g reagent is designed for detecting glucuronic acid and β-glucuronidase activity in biological samples. It produces a colorimetric response when the enzyme breaks down the substrate, providing a simple and effective means of measuring enzyme activity. The compound is widely used in clinical laboratories, research facilities, and diagnostic applications, making it an essential tool for understanding glucuronidation processes and related enzyme functions in various biological systems.
Catalog No. 50-006-61772