Learn More
Alkali Scientific X GlcA Cyclohexylammonium Salt [5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt], 250 Mg
SDP
Supplier:  Alkali Scientific XGL003CP
Encompass_Preferred
X GlcA Cyclohexylammonium Salt (5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt) is a highly sensitive chromogenic substrate used in laboratory assays to detect the presence of glucuronic acid in biological samples. This 250 mg reagent is commonly used in enzymatic assays, including those for β-glucuronidase, to assess enzyme activity. The product produces a colorimetric change, allowing researchers to monitor enzyme reactions visually. This substrate is ideal for applications in biochemical research, diagnostics, and clinical assays where precise detection and quantification of glucuronic acid derivatives are required.
Catalog No. 50-006-60599