Learn More
Electron Microscopy Sciences X-Ray Window Silicon Dioxide G-FLAT, 500 µm, 100 nm
SDP
Supplier:  Electron Microscopy Sciences 7604210
Encompass_Preferred
Ideal substrates for x-ray microscopy and x-ray spectroscopy techniques. State-of-the-art manufacturing and expert engineering allow for competitive prices of these windows. Made in the USA, these X-Ray windows are entirely plasma cleanable to remove organic contamination. Flat, uniformly deposited films provide consistent backgrounds with low field-to-field variability and high x-ray transmission. For biological imaging studies, with a glass-like hydrophilic surface.
Dimensions: 500 µm
Catalog No. 50-192-9440
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.