Learn More
Electron Microscopy Sciences Tripod Polisher X-Section L-Bracket
SDP
Supplier:  Electron Microscopy Sciences 5011631
Encompass
X-Section L-Bracket (#50116-31): is used to mechanically clamp a sample (such as an IC Package) on the side of an L-Bracket for optimal SEM cross sectioning. - For use with Tripod Polisher Model 590.
Catalog No. 50-980-870