Learn More
Leica Biosystems X-tra Adhesive White (1440)
Supplier:  Leica Biosystems 3800200AE
Encompass_Preferred
X-tra Slides have a positive charged surface which helps to bond tissue sections and cytology preparations.X‐tra slides are used for demanding H&E procedures that require tissue adhesion. Size: 1” x 3” x .04” (unless otherwise noted)Hydrophobic surface with approximately 90 degree contact angle. Recommended uses include special stains, IHC, ISH, cryosectioning, and cytology.
Catalog No. 50-255-2449