Learn More
Ted Pella Inc PELCO X-TREME SEM Storage Box and holder for up to 18 of the 12.7mm SEM pin mounts
SDP
Supplier:  Ted Pella Inc 16799
Encompass_Preferred
Very strong and rugged clear polycarbonate box to store standard pin mounts with 3.2mm (1/8"). This box is watertight, crushproof and dustproof. The strong hinges, locking clasps and soft silicone O-ring seal makes this an ideal transport, shipping and storage box for SEM samples on pin mounts. The holder insert is numbered 1-18 and accommodates all types of pin mounts form 6.4 to 50mm diameter.
Catalog No. NC1958580