Learn More
Fluidigm Corp X8 ANTIBODY LABELING 4RXN
Supplier: Fluidigm Corp 201173A
This Maxpar kit lets you conjugate IgG antibodies with the 173Yb metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.