Learn More
ATCC X9; Fibroblast; Mouse (Mus musculus)
Supplier:  ATCC CRL3282
Encompass
X9 is a fat cell line that was isolated in 2010 from the inguinal subcutaneous fat depot of a 7-to-90-week mouse. This cell line was deposited by BM Spiegelman.
Catalog No. 50-238-2509