Learn More
Medchemexpress LLC Xaliproden hydrochloride | 90494-79-4 | >96.0% | C24H23ClF3N | 10MG
SDP
Supplier:  Medchemexpress LLC HY1460410MG
Encompass_Preferred
Xaliproden hydrochloride is the hydrochloride salt of xaliproden, an orally active, non-peptidic 5-hydroxytryptamine (5-HT) 1A receptor agonist with reported neurotrophic and neuroprotective activity. Supplied as a research-grade solid, it is intended for in vitro and in vivo studies of 5-HT1A-mediated signaling and neuroprotection. Not for human or diagnostic use.
  • Potent 5-HT1A receptor agonist.
  • Shows neurotrophic and neuroprotective activity.
  • Suitable for in vitro and in vivo pharmacology research.
  • Typical purity ≥96.0%.
  • Molecular weight 417.9 g/mol; formula C24H23ClF3N.
  • Provided as a solid research reagent; consult CoA for lot-specific data.
Catalog No. 50-002-58579