Learn More
Apexbio Technology LLC Xamoterol hemifumarate 73210-73-8 25mg
SDP
Supplier:  Apexbio Technology LLC B651925
Encompass_Preferred
Xamoterol hemifumarate (CAS 73210-73-8) is a selective 1-adrenergic receptor agonist targeting cardiac 1 receptors It is designed to activate 1-adrenergic signaling pathways thereby modulating myocardial function and cardiac electrophysiological processes Xamoterol hemifumarate exerts its biological activity primarily through 1-adrenergic receptor stimulation influencing downstream intracellular signaling cascades related to cardiac rhythm regulation In experimental settings it is utilized in vitro with cellular cardiac models and in vivo with animal models to assess 1-adrenergic receptor-mediated effects particularly in the interaction with cardiac ion channels such as the rapid delayed rectifier potassium current (IKr) Based on these pharmacological properties Xamoterol hemifumarate holds research potential in the study of arrhythmogenesis mechanisms and cardiac electrophysiology
Catalog No. 50-000-92035