Learn More
Medchemexpress LLC Xamoterol hemifumarate | 73210-73-8 | MFCD00661103 | 99.9% | 397.43 | C20H29N3O9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101327A10MG
Encompass_Preferred
Xamoterol hemifumarate is a research-grade β1-adrenergic receptor agonist provided as the hemifumarate salt for in vitro and in vivo pharmacology and cardiac electrophysiology studies.
  • Selective β1-adrenergic receptor agonist activity
  • Supplied as a stable solid hemifumarate salt
  • High purity (99.92%) appropriate for research use
  • Defined chemical identity: CAS 73210-73-8; MW 397.43
  • Available in small pack sizes for laboratory experiments
Catalog No. 50-002-12814