Learn More
Medchemexpress LLC Xamoterol hemifumarate | 73210-73-8 | MFCD00661103 | 99.9% | 397.43 g/mol | C20H29N3O9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101327A5MG
Encompass_Preferred
Xamoterol hemifumarate is the hemifumarate salt of xamoterol, a selective beta1-adrenergic receptor agonist used as a research reagent for studies of adrenergic signaling and cardiac arrhythmogenesis. It is supplied as a high-purity solid with options for small-scale mg quantities or DMSO solutions for in vitro assays.
  • Selective beta1-adrenergic receptor agonist suitable for receptor pharmacology studies.
  • High purity (>99.9%) suitable for analytical and biological assays.
  • Supplied as a solid, stable for storage and formulation.
  • Available in small mg quantities and as DMSO solutions for immediate use.
  • Molecular weight 397.43 g/mol; CAS 73210-73-8.
  • Useful for studies of cardiac electrophysiology and arrhythmogenesis.
Catalog No. 50-002-12923