Learn More
Medchemexpress LLC Xamoterol hemifumarate | 73210-73-8 | MFCD00661104 | >98.0% | 397.43 | C20H29N3O9 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101327A25MG
Encompass_Preferred
Xamoterol hemifumarate is the hemifumarate salt of xamoterol, a selective β1-adrenergic receptor agonist used in cardiac electrophysiology and receptor pharmacology research. It is supplied as a pre-weighed research chemical suitable for in vitro assays. CAS: 73210-73-8; formula: C20H29N3O9; molecular weight: 397.43.
Catalog No. 50-002-11265