Learn More
Selleck Chemical LLC Xanomeline E4714-1G
SDP
Supplier:  Selleck Chemical LLC E47141G
Encompass_Preferred
Xanomeline(LY-246708) is a selective agonist of muscarinic type 1(M(1) receptor and muscarinic type 4 receptor(M(4) It increases neuronal excitability and inhibits behaviors induced by D-amphetamine and (-)-apomorphine
Catalog No. 50-000-39560